Order Entry
Denmark
ContactUsLinkComponent
(R)-4-(4-Methoxybenzyloxy)-1,2-butanediol ≥96.0% (by GC)
(R)-4-(4-Methoxybenzyloxy)-1,2-butanediol ≥96.0% (by GC)
Catalog # TCIAM1788-500MG
Supplier:  TCI
CAS Number:  
(R)-4-(4-Methoxybenzyloxy)-1,2-butanediol ≥96.0% (by GC)
Catalog # TCIAM1788-500MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Specific rotation [a]20/D:
    -6.0 to -10.0 deg (C=1, CHCl3)

Specification Test Results

About this item

 :
 :226.27 g/mol
CAS Number:213978-61-1
Formula:C₁₂H₁₈O₄