Methyl-3,4,6-tri-O-acetyl-2-deoxy-2-phthalimido-1-thio-β-D-glucopyranoside ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:79528-48-6
:
Formula:C₂₁H₂₃NO₉S
:465.47 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:148.0 to 152.0 deg-C
- Specific rotation [a]20/D:+48.0 to +51.0 deg (C=1, CHCl3)
Specification Test Results
Frequently Bought Together