Order Entry
Denmark
ContactUsLinkComponent
cis-β-Methylstyrene ≥98.0% (by GC) stabilised
Flammable
Irritant
cis-β-Methylstyrene ≥98.0% (by GC) stabilised
Catalog # TCIAM1174-10ML
Supplier:  TCI
CAS Number:  
Flammable
Irritant
cis-β-Methylstyrene ≥98.0% (by GC) stabilised
Catalog # TCIAM1174-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :118.18 g/mol
Boiling Pt:174°C
Melting Pt:-60°C
Melting Pt:–60°C
Flash Pt:45°C
UN:3295
ADR:3,III
 :Stabilised with 4-tert-butylpyrocatechol (TBC)
 :GHS02
 :GHS07
CAS Number:766-90-5
Formula:C₉H₁₀
MDL Number:MFCD00065137