(2-Methoxyethoxymethyl)triethylammonium chloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAM0681-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:62.0 to 66.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:225.76 g/mol
CAS Number:60043-43-8
Formula:C₁₀H₂₄ClNO₂
MDL Number:MFCD00059973