(-)-ɑ-Phellandrene >65.0% (by GC)
Supplier: TCI
About this item
Synonyms: (-)-p-Mentha-1,5-diene, (-)-5-Isopropyl-2-methyl-1,3-cyclohexadiene, (-)-α-Phellandrene, ['(-)-5-Isopropyl-2-methyl-1,3-cyclohexadiene', '(-)-', 'p', '-Mentha-1,5-diene']
- CAS Number:4221-98-1
- Formula:C₁₀H₁₆
- MDL Number:MFCD00062988
- Molecular Weight:136.24 g/mol
- Merck Index:14,07200
- Boiling Pt:175°C
- Flash Pt:49°C
- Storage Temperature:Refrigerator
- EINECS:224-167-6
- UN:2319
- ADR:3,III
2 Options Available Below
Product Details & Documents
Synonyms: (-)-p-Mentha-1,5-diene, (-)-5-Isopropyl-2-methyl-1,3-cyclohexadiene, (-)-α-Phellandrene, ['(-)-5-Isopropyl-2-methyl-1,3-cyclohexadiene', '(-)-', 'p', '-Mentha-1,5-diene']
- CAS Number:4221-98-1
- Formula:C₁₀H₁₆
- MDL Number:MFCD00062988
- Molecular Weight:136.24 g/mol
- Merck Index:14,07200
- Boiling Pt:175°C
- Flash Pt:49°C
- Storage Temperature:Refrigerator
- EINECS:224-167-6
- UN:2319
- ADR:3,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 65.0 %
- Specific rotation [a]20/D:-140.0 to -190.0 deg (neat)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



