Order Entry
Denmark
ContactUsLinkComponent
Lead(II) acetate trihydrate ≥99.0% (by titrimetric analysis)
Corrosive
Health
Environment
Lead(II) acetate trihydrate ≥99.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
Health
Environment
Lead(II) acetate trihydrate ≥99.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Chelometric Titration):
    min. 99.0 %

Specification Test Results

Related Information

Frequently Bought Together