Order Entry
Denmark
ContactUsLinkComponent
Lithium bromide ≥99.0% (by titrimetric analysis)
Irritant
Lithium bromide ≥99.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Lithium bromide ≥99.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity (Argentmetric Titration):
    min. 99.0 %

Specification Test Results

Frequently Bought Together