Lectin, fucose specific 5 mg/mL in PBS (from Aspergillus oryzae)
TCIAL0169-1ML
: TCI
:
- Pk:1 mL
- Seq:2070
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Concentration (Lowry Methoed):4.4 to 5.5 mg/mL
- Molecular weight:30000 ~ 40000 (SDS-PAGE)
- Hemagglutination activity:max. 15.6 micro g/mL (pH6.5)
:
:Refrigerator
:MFCD06798677