Order Entry
Denmark
ContactUsLinkComponent
2-Methylpropene ∼8% in dichloromethane
2-Methylpropene ∼8% in dichloromethane
Supplier:  TCI
Certificates
undefined
2-Methylpropene ∼8% in dichloromethane
Supplier:  TCI
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (HNMR):
    8.0 to 9.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together