Imperatorin ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:482-44-0
:
Formula:C₁₆H₁₄O₄
MDL Number:MFCD00016881
:270.284 g/mol
Merck Index:14,04925
Storage Temperature:Freezer
EINECS:207-581-1
Specification Test Results
- Purity (HPLC):min. 98.0 area%
- Melting point:97.0 to 101.0 deg-C
Specification Test Results
Frequently Bought Together