Order Entry
Denmark
ContactUsLinkComponent
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Irritant
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Catalog # TCIAI0873-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-Isobutylcyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Catalog # TCIAI0873-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 % (total of isomers)
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :184.28 g/mol
 :GHS07
CAS Number:38792-88-0
Formula:C₁₁H₂₀O₂