(S)-(-)-Sulpiride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:23672-07-3
:
Formula:C₁₅H₂₃N₃O₄S
MDL Number:MFCD00055220
:341.43 g/mol
Merck Index:14,08989
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:185.0 to 189.0 deg-C
- Specific rotation [a]20/D:-67.0 to -71.0 deg (C=3, DMF)
Specification Test Results
Frequently Bought Together