
(2-Hydroxyethyl)dimethyl(3-sulphopropyl)ammonium hydroxide inner salt ≥98.0% (by HPLC, total nitrogen) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:38880-58-9
:
Formula:C₇H₁₇NO₄S
MDL Number:MFCD01320397
:211.28 g/mol
EINECS:254-176-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:246.0 to 250.0 deg-C
- Suitability for protein-analysis:to pass test
Specification Test Results
Frequently Bought Together