Biotin-LC-hydrazide ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:109276-34-8
:
Formula:C₁₆H₂₉N₅O₃S
MDL Number:MFCD00077659
:371.50 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):18.00 to 19.50 %
Specification Test Results
Frequently Bought Together