Order Entry
Denmark
ContactUsLinkComponent
 
2-(Hydroxymethyl)-2-nitro-1,3-propanediol ≥99.0% (by GC)
 :  TCI
 
2-(Hydroxymethyl)-2-nitro-1,3-propanediol ≥99.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Solubility in Water:
    almost transparency

 

Frequently Bought Together