Gomisin C ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:58546-56-8
:
Formula:C₃₀H₃₂O₉
MDL Number:MFCD09026937
:536.58 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together