GlcNAc beta(1-4)[Fuc alpha(1-6)]GlcNAc ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:108964-40-5
:
Formula:C₂₂H₃₈N₂O₁₅
:570.55 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 98.0 area%
Specification Test Results
Frequently Bought Together