
2-Furohydrazide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAF1015-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:77.0 to 81.0 deg-C
Specification Test Results
About this item
:
:126.11 g/mol
Boiling Pt:279°C
EINECS:222-046-2
:GHS07
CAS Number:3326-71-4
Formula:C₅H₆N₂O₂
MDL Number:MFCD00003235