
4-Fluorobenzamidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAF0959-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Melting point:209.0 to 213.0 deg-C
- Water:15.0 to 20.0 %
Specification Test Results
About this item
:
:174.61 g/mol
:GHS07
CAS Number:456-14-4
Formula:C₇H₈ClFN₂
MDL Number:MFCD04114421