5-Formamido-1-[2-(formyloxy)ethyl]pyrazole ≥97.0% (by GC, titration analysis)
Catalog # TCIAF0890-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:84.0 to 88.0 deg-C
Specification Test Results
About this item
:
:183.17 g/mol
CAS Number:116856-18-9
Formula:C₇H₉N₃O₃
MDL Number:MFCD09744040