Fmoc-α-methylalanine ≥98.0% (by HPLC, titration analysis)
TCIAF0888-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
: N-Fmoc-alpha-methylalanine, 2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]isobutyric acid, Fmoc-Aib-OH, 2-(Fmoc-amino)isobutyric acid
:
:325.364 g/mol
:Refrigerator
:94744-50-0
:C₁₉H₁₉NO₄
:MFCD00151913