

Fluorescein isothiocyanate (mixture of 5 and 6 isomers) ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:27072-45-3
:
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063
Merck Index:14,04089
:389.38 g/mol
EINECS:248-207-7
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 97.0 % (total of isomer)
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together