
4-Fluoro-1,3-dioxolan-2-one ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:114435-02-8
:
Formula:C₃H₃FO₃
MDL Number:MFCD06247543
:106.05 g/mol
Boiling Pt:210°C
Melting Pt:20°C
Storage Temperature:Refrigerator
EINECS:483-360-5
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together