N-Fmoc-L-alanine ≥98.0% (by HPLC, titration analysis)
: TCI
: N-(9-Fluorenylmethoxycarbonyl)-L-alanine, (-)-N-{[(9H-Fluoren-9-yl)methoxy]carbonyl}-L-alanine, Fmoc-L-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-alanine, (-)-L-Alanine, N-FMOC protected, (-)-FMOC-L-Ala-OH, (2S)-(-)-2-Aminopropanoic acid, N-FMOC protected, Fmoc-Ala-OH
:35661-39-3
:
:C₁₈H₁₇NO₄
:MFCD00037139
:311.34 g/mol
:252-660-6
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:-17.0 to -21.0 deg (C=1, DMF)
- Solubility in Methanol:almost transparency
Frequently Bought Together