

FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3326-32-7
:
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063
:389.381 g/mol
EINECS:222-042-0
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together