Order Entry
Denmark
ContactUsLinkComponent
 
(3,4-Epoxycyclohexyl)methyl acrylate ≥94.0% (by GC) stabilised
 :  TCI
 
(3,4-Epoxycyclohexyl)methyl acrylate ≥94.0% (by GC) stabilised
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 94.0 %

 

Frequently Bought Together