Order Entry
Denmark
ContactUsLinkComponent
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate ≥98.0% (by GC)
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate ≥98.0% (by GC)
Supplier:  TCI
Certificates
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    36.0 to 40.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together