Order Entry
Denmark
ContactUsLinkComponent
4-Ethynyl-N,N-dimethylaniline ≥98.0% (by GC, titration analysis)
Irritant
4-Ethynyl-N,N-dimethylaniline ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
4-Ethynyl-N,N-dimethylaniline ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    49.0 to 53.0 deg-C

Specification Test Results

Frequently Bought Together