Order Entry
Denmark
ContactUsLinkComponent
3-Ethyl-3-methylpentane ≥99.0% (by GC)
 
 
 
 
3-Ethyl-3-methylpentane ≥99.0% (by GC)
  TCIAE0258-5ML
 :  TCI
 :  
 
 
 
 
3-Ethyl-3-methylpentane ≥99.0% (by GC)
  TCIAE0258-5ML
 :  TCI
 :  

 

  • Pk:
    5 mL
  • Seq:
    2210

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :114.23 g/mol
 :118°C
 :6°C
 :213-923-0
 :1262
 :3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
 :1067-08-9
 :C₈H₁₈
 :MFCD00048630