5-Methyl-3-hexanone ≥98.0% (by GC)
Catalog # TCIAE0249-10ML
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:114.19 g/mol
Boiling Pt:136°C
Flash Pt:39°C
EINECS:210-801-9
UN:1224
ADR:3,III
:GHS02
CAS Number:623-56-3
Formula:C₇H₁₄O
MDL Number:MFCD00009319