

EDTA (ethylenediamine tetraacetic acid) ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:60-00-4
:
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541
:292.25 g/mol
Merck Index:14,03517
Melting Pt:242°C
EINECS:200-449-4
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (Chelometric Titration):min. 98.0 %
- Solubility in 1mol/L NaOH:almost transparency
Specification Test Results
Frequently Bought Together