Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dextrin, Soluble fiber

Dextrin, Soluble fiber

Supplier:   TCI
Dextrin, Soluble fiber
Supplier:  TCI
Certifikater

2 tilgængelige muligheder nedenfor

Product Details & Documents
Synonymer:  Dextrin
  • CAS nummer:9004-53-9
  • Formel:(C₆H₁₂O₆)n
  • MDL nummer:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,02953
  • EINECS:232-675-4
Specifications for Product Family

Specifikationstestresultater

  • Appearance:
    White to Almost white powder to crystal
  • Solubility in hot Water:
    transparency
  • Drying loss:
    max. 10.000 %
  • Ignition residue (Sulfate):
    max. 0.5 %
  • Reducing suger:
    max. 8.0
Product Family Options

Produktinformation

  • Pk
    Availability
    Price
  • 100 g/mol
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 100 g/mol
    1 * 100 g/mol - 132,00 kr
    132,00 kr
    1 * 100 g/mol
       132,00 kr

  • 500 g/mol
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 g/mol
    1 * 500 g/mol - 362,00 kr
    362,00 kr
    1 * 500 g/mol
       362,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.