
N,N-Dimethyl-1,3-propanediamine dihydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:52198-63-7
:
Formula:C₅H₁₄N₂·₂HCl
MDL Number:MFCD32062892
:175.10 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:187.0 to 191.0 deg-C
Specification Test Results
Frequently Bought Together