
2-(Diphenylphosphino)benzonitrile ≥98.0% (by GC, titration analysis)
TCIAD4953-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Iodometric Titration):min. 98.0 %
- Melting point:148.0 to 152.0 deg-C
:
:287.30 g/mol
:GHS07
:34825-99-5
:C₁₉H₁₄NP
:MFCD05864011