N,N-Dimethylisobutyramide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:21678-37-5
:
Formula:C₆H₁₃NO
MDL Number:MFCD00075378
:115.18 g/mol
Boiling Pt:179°C
Flash Pt:70°C
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together