Order Entry
Denmark
ContactUsLinkComponent
7H-Pyrrolo[2,3-d]pyrimidine ≥98.0% (by GC, titration analysis)
Irritant
7H-Pyrrolo[2,3-d]pyrimidine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
7H-Pyrrolo[2,3-d]pyrimidine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    135.0 to 139.0 deg-C

Specification Test Results

Frequently Bought Together