Dicamba ≥98.0% (by GC, titration analysis)
TCIAD4800-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:113.0 to 117.0 deg-C
: 3,6-Dichloro-2-methoxybenzoic acid, MDBA, 3,6-Dichloro-o-anisic acid
:
:221.03 g/mol
:14,03040
:217-635-6
:GHS05
:GHS07
:Refrigerator
:1918-00-9
:C₈H₆Cl₂O₃
:MFCD00055283