
2,4-Dihydroxybenzhydrazide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD4784-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:168.15 g/mol
:GHS07
CAS Number:13221-86-8
Formula:C₇H₈N₂O₃
MDL Number:MFCD00017053