Order Entry
Denmark
ContactUsLinkComponent
2,2-Difluoroethylamine ≥98.0% (by GC, titration analysis)
2,2-Difluoroethylamine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
2,2-Difluoroethylamine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together