Diethyl-trans-1,2-cyclopropanedicarboxylate ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3999-55-1
:
Formula:C₉H₁₄O₄
MDL Number:MFCD00198002
:186.207 g/mol
Boiling Pt:71°C
Flash Pt:98°C
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together