Diethylene glycol isopropyl methyl ether ≥98.0% (by GC)
Catalog # TCIAD4677-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
Synonyms: 2-[2-(2-Methoxyethoxy)ethoxy]propane
:
:162.23 g/mol
CAS Number:77078-22-9
Formula:C₈H₁₈O₃
MDL Number:MFCD28009631