2,5-Diaminotoluene ≥98.0% (by GC, titration analysis)
: TCI
: 2-Methyl-1,4-phenylenediamine
:95-70-5
:
:C₇H₁₀N₂
:MFCD00035779
:168°C
:122.17 g/mol
:202-442-1
:2811
:6.1 III
:GHS06
:GHS09
- Environmentally Preferable:
- Appearance:White to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C
Frequently Bought Together