3,3,3',3'-Tetramethyl-1,1'-bis(4-sulphobutyl)indocarbocyanine sodium salt ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD4464-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Yellow to Dark green to Black powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 % (calcd.on anh.substance)
- Lambda max.:547.0 to 552.0 nm (MeOH)
- Absorbance (E1%1cm):min. 2000 (MeOH, 547.0 to 552.0 nm)
Specification Test Results
About this item
:
:622.77 g/mol
CAS Number:120724-84-7
Formula:C₃₁H₄₀N₂O₆S₂
MDL Number:MFCD16621066