Order Entry
Denmark
ContactUsLinkComponent
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10) ≥98.0% (by HPLC, total nitrogen) for biochemical research
Irritant
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10) ≥98.0% (by HPLC, total nitrogen) for biochemical research
Supplier:  TCI
Certificates
Irritant
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10) ≥98.0% (by HPLC, total nitrogen) for biochemical research
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Suitability for protein-analysis:
    to pass test

Specification Test Results

Related Information

Frequently Bought Together