N,N-Diisopropylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
: TCI
: Diisopropylcarbamoyl Chloride
:19009-39-3
:
:C₇H₁₄ClNO
:MFCD00015013
:100°C
:163.6452 g/mol
:242-743-5
:3261
:8 II
:GHS05
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:57.0 to 61.0 deg-C
Frequently Bought Together