
(S)-(-)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:848821-58-9
:
Formula:C₂₀H₂₇NOSi
MDL Number:MFCD08690083
:325.53 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:-48.0 to -53.0 deg (C=1, CHCl3)
Specification Test Results
Frequently Bought Together