2,6-Dibromodithieno[3,2-b:2',3'-d]thiophene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:67061-69-2
:
Formula:C₈H₂Br₂S₃
MDL Number:MFCD11215199
:354.09 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:168.0 to 174.0 deg-C
Specification Test Results
Frequently Bought Together