


Dichloromethane-[D₂]
Catalog # TCIAD3529-10ML
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Degree of deuteration:min. 99.9 atom%D
Specification Test Results
About this item
:
:86.92 g/mol
EINECS:216-776-0
UN:1593
ADR:6.1,III
:GHS07
:GHS08
:GHS09
CAS Number:1665-00-5
Formula:CD₂Cl₂
MDL Number:MFCD00000882