2,6-Dibromo-1,5-dihydroxynaphthalene ≥93.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:84-59-3
:
Formula:C₁₀H₆Br₂O₂
MDL Number:MFCD00021473
:317.96 g/mol
EINECS:201-543-8
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 93.0 %
Specification Test Results
Frequently Bought Together