
4-Dodecylbenzenesulphonyl azide mixture, soft type
Catalog # TCIAD2580-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Lambda max.:234.0 to 237.0 nm (CH3CN)
- Absorbance (E1%1cm):min. 400 (CH3CN, 234.0 to 237.0 nm)
Specification Test Results
About this item
:
:351.51 g/mol
:GHS07
CAS Number:79791-38-1
Formula:C₁₈H₂₉N₃O₂S
MDL Number:MFCD00274161